Learn More
Sigma Aldrich m-Phenylenediamine dihydrochloride
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- M-phenylenediamine dihydrochloride is used in the synthesis of bis(thiazolidone) and bis(thiohydantoin) derivatives
- It can be used to increase the pore size in the fabrication of poly(aniline-co-1,3-phenylenediamine) cryogels
Specifications
Specifications
| CAS | 541-69-5 |
| Molecular Formula | C6H8N2 -+ 2 HCl |
| Linear Formula | C6H8N2 -+ 2 HCl |
| MDL Number | MFCD00012975 |
| Quantity | 100 g |
| Synonym | 1,3-Benzenediamine dihydrochloride |
| Molecular Weight (g/mol) | 181.06 |
| Percent Purity | ≥99.0% (AT) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.