Learn More
Branson Ultrasonics™ M Series Ultrasonic Cleaning Bath
Outfit your lab with a Bransonic MH Series Ultrasonic Cleaner. This unit is designed with solid construction and excellent functionality for a variety of applications − including cell separation, sample preparation and degassing of liquids.
Supplier:  Branson Ultrasonics™ CPX952536R
 View more versions of this product
Catalog No. 15-336-108
Only null left
Add to Cart
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Description
Outfit your lab with a Bransonic MH Series Ultrasonic Cleaner. This unit is designed with solid construction and excellent functionality for a variety of applications − including cell separation, sample preparation and degassing of liquids.
  • Sweep frequency eliminates standing waves and creates consistent cavitation throughout the tank, eliminating dead zones for long life
  • Includes 60-minute mechanical timer and continuous operation
  • 40kHz industrial transducers
  • Controls located above and behind in easily accessible control tower panel which is raised above tank to avoid damage from cleaning solutions
  • Plastic body
  • Chemical-resistant
  • 1.5 gal. and larger models
  • Tank drains with valves
Specifications
Specifications
None - Control Dial
Plastic Housing, Stainless Steel Tank
230/240 V
9.5 L
Tank cover
14 lb.
2 Years
60 min.
40 kHz
2.5 gal.
29 x 24 x 15 cm (11.5 x 9.5 x 6 in.)
M5800-E
6.4 kg
CEE 7/7 (Continental European)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.