Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Tolyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. I045925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I045925G 25 g
I04595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I045925G Supplier TCI America Supplier No. I045925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 621-30-7
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
MDL Number MFCD00004807
InChI Key BDPQUWSFKCFOST-UHFFFAOYSA-N
Synonym m-tolyl isothiocyanate, 3-methylphenyl isothiocyanate, m-tolylisothiocyanate, benzene, 1-isothiocyanato-3-methyl, 3-tolyl isothiocyanate, isothiocyanic acid m-tolyl ester, 1-isothiocyanato-3-methyl-benzene, 1-isothiocyanato-3-methylbenzol, 3-isothiocyanatotoluene, acmc-209mzt
PubChem CID 69304
IUPAC Name 1-isothiocyanato-3-methylbenzene
SMILES CC1=CC=CC(=C1)N=C=S

Specifications

Color Yellow
Boiling Point 240°C
Quantity 25 g
UN Number 2810
Formula Weight 149.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material m-Tolyl Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.