Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylene, 99%, Extra Dry, AcroSeal™

Catalog No. p-4626817
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC158931000 100 mL
AC158930010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158931000 Supplier Thermo Scientific Chemicals Supplier No. 158931000
Only null left
Add to Cart

CAS: 108-38-3 | C8H10 | 106.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-38-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008536
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150
PubChem CID 7929
ChEBI CHEBI:28488
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -48.0°C
Appearance clear colorless liquid
Density 0.8640g/mL
Quantity 100 mL
Flash Point 25°C
Refractive Index 1.4960 to 1.4980
Assay Percent Range 99%
Specific Gravity 0.864
Linear Formula C6H4(CH3)2
Viscosity 0.62 mPa.s (20°C)
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
Boiling Point 139.0°C
Formula Weight 106.17
Percent Purity 99%
Grade Extra Dry
Chemical Name or Material m-Xylene
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation
Harmful in contact with skin
Harmful if inhaled
Flammable liquid and vapor
Precautionary Statement WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-576-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.