Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-MDA-D5 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco M0271ML

Catalog No. 01-001-581


Only null left
Add to Cart

Description

Description

A stable-labeled internal standard suitable for quantitation of MDA levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. MDA, or 3,4-methylenedioxyamphetamine, is an illicit recreational drug of the amphetamine and phenethylamine classes. Also known as tenamphetamine, MDA is abused for its psychedelic and hallucinogenic effects, similar to those of MDMA or "ecstasy".
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1, 136765-42-9
MFCD00673286
1.0 mg/mL in methanol
184.25
1 mL
C10D5H8NO2
UN 1230 - class 3 - PG 2 - Methanol, solution
(+/-)-3,4-Methylenedioxyamphetamine-d5
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

CERILLIANT is a registered trademark of Cerilliant Corporation