Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ M12 Male to M12 Male Communications Cable

Catalog No. 19029958 Shop All Brady Corporation Products
Change view
Click to view available options
Length (English):
16.4 ft.
32.81 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-029-958 16.4 ft.
19-029-959 32.81 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-029-958 Supplier Brady™ Supplier No. 178333
Add to Cart
Add to Cart

For use with the V1400 Industrial Ethernet Bluetooth Cradle.

M12 Male to M12 Male Communications cable for use with V1400 Industrial Ethernet Bluetooth Cradele. Available in 5 m or 10 m lengths.

Specifications

Length (English) 16.4 ft.
Length (Metric) 5 m
Type Cable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.