Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ M12 Ultra™ Electrophoresis and Visualization Package
SDPSCI_ED

Catalog No. S99498 Shop All EDVOTEK Products
Change view
Click to view available options
Gel Size:
7 x 7 cm, 7 x 14 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gel Size
S99498 7 x 7 cm, 7 x 14 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S99498 Supplier Edvotek™ Supplier No. 505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The M12 Ultra™ Kit includes the M12 Complete™ Electrophoresis Package and the all-new TruBlu™ Jr. Blue Light Transilluminator. This dynamic duo enables real-time visualization of SYBR™ Safe or other blue light-reactive dye agarose gels.

The M12 Ultra™ offers ultimate flexibility, allowing you to either visualize gels in real-time with the orange contrast lid and TruBlu™ Jr., or to stain your gels post-electrophoresis if you plan to use a traditional blue stain. This package supports one or two student groups in two standard length gel trays for experiments that require less separation, or one long gel tray for experiments that require more. Produces excellent results in 15 to 30 minutes.

For blue light-reactive gels, overlay the orange filter on the M12 lid and slide the TruBlu™ Jr. underneath for instant visualization! The TruBlu™ Jr. can also be used to examine fluorescence from green fluorescent protein (GFP).

TruBlu™ Jr. features:

  • Battery operated for convenient use in the classroom
  • Designed to work seamlessly with the Edvotek™ M12 Electrophoresis System
  • Includes an orange contrast lid
  • Safe, non-UV wavelength
  • Blue light emission spectrum centered around 470 nm is compatible with SYBR™ Safe, GelGreen™ and other blue light-reactive dyes
  • 10 x 6 cm viewing surface

M12 Complete™ features:

  • Sleek new design speeds electrophoresis
  • Includes complete set of new and improved electrophoresis accessories
  • Now includes a DNA DuraGel™ for pipetting practice
  • Large, color-coded push tabs for easy lid insertion and removal
  • Pour spout for buffer disposal
  • Improved ventilation reduces lid condensation
  • User replaceable electrodes
  • Chamber reverse compatible with previous Edvotek™ accessories

Specifications

Capacity 2 Gel Trays
Detection Method LED blue
For Use With (Application) Gel Electrophoresis
For Use With (Equipment) Pipette, Power Supply
Gel Size 7 x 7 cm, 7 x 14 cm
Max. Current 500 mA
Product Line Edvotek
Running Time 15 to 30 min.
Timer 0 to 99 min. or continuous
Type Electrophoresis Kit
Includes (2) 7 x 7 cm Gel Trays, 7 x 14 cm Gel Tray, (2) 6/8 Tooth Combs, (4) Rubber End Caps, DNA DuraGel™, TruBlu™ Jr., Orange Contrast Lid
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.