Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ M13 Human, Forward Primer, HPLC

Catalog No. A15635
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15635 50 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15635 Supplier Applied Biosystems™ Supplier No. A15635
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M13 Human, Forward Primer, HPLC

Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.

TRUSTED_SUSTAINABILITY

Specifications

Description M13 Human Forward HPLC Primer, 50 nmole, For PCR and Sanger Sequencing
Product Type M13 Human Forward HPLC Primer
Primer Forward
Quantity 50 nmol
Grade HPLC
I am trying to use the Expi293 Expression System in 96-well microtiter plates and am not getting good yields. Do you have any tips for optimizing in this format?

Please see the Application Note (http://www.thermofisher.com/content/dam/LifeTech/migration/files/proteins-expression-isolation-analysis/pdfs.par.28891.file.dat/expi293-microtiter-plates-co25793.pdf) for tips on optimizing protein yield using the Expi293 Expression System in 96-well microtiter plates.

I am getting very poor yield of protein using the Expi293 Expression System. Do you have any suggestions for optimizing protein expression?

Please refer to the Application Note (http://www.thermofisher.com/content/dam/LifeTech/migration/files/proteins-expression-isolation-analysis/pdfs.par.61153.file.dat/expi293-protein-yield-co25763.pdf for tips on optimizing protein yield using the Expi293 Expression System.

What should I do if my predesigned primers ordered using the Primer Designer Tool do not work due to no amplification, off‐target amplification, or SNP of interest being too close to the PCR priming region?

If there are issues with the predesigned primers ordered via the Primer Designer Tool, please contact Thermo Fisher Technical Support (https://www.thermofisher.com/us/en/home/technical-resources/contact-us.html).

Who can I contact if I do not receive my primers after ordering them using the Primer Designer Tool?

To find more information about the status of the primers ordered using the Primer Designer Tool, please contact Thermo Fisher Scientific Product Support (https://www.thermofisher.com/us/en/home/technical-resources/contact-us.html) and have your order number available.

How can I obtain the amplicon sequence for a primer pair when using the Primer Designer Tool?

In the Primer Designer Tool, the amplicon coordinates are displayed for each primer pair. At this time, we do not provide the amplicon sequences from our webpage. To obtain the amplicon sequence, we recommend the NCBI website. You can click on View Details within the Primer Designer Tool and there are several hyperlinks that will take you to different locations on the NCBI page. For example, if you click on the Entrez Gene ID hyperlink, it will take you to the Gene ID page. You can scroll down to Genomic regions, transcripts, and products and click on the FASTA option next to "Go to nucleotide" and that will bring up the whole sequence.

In the Primer Designer Tool, how large is the intron span on either end for the predesigned primers?

In the Primer Designer Tool, many of the primer pairs span the entire exon, and hence provide some sequence for exon adjacent sequence. The extent of this intronic sequence varies from assay to assay and the extent can be viewed in the genome browser panel. Currently, we only provide primers for exons and are not offering "intron only" amplicon primers.

Does the Primer Designer Tool design primers in exon regions?

The primer pairs obtained via the Primer Designer Tool amplify exon regions of genes. In order to have complete coverage, the first and last primer of each exon typically lies in an intron. There may be some instances where primers are designed in an exon region, due to the size of the exon.

When using the Primer Designer Tool, what should the distance from a SNP of interest be relative to the 3' end of the primer?

When using the Primer Designer Tool, the suggested distance of the SNP of interest relative to the 3' end of the primer depends upon the BigDye sequencing kit being used, the use of M13 tails, and the polymer type. BigDye Direct and BigDye Terminator v1.1 typically have higher data quality very close to the sequencing primer end, whereas BigDye Terminator v3.1 together with POP‐7 polymer provides optimized data quality after the first 35 to 40 bases. Predesigned primers ordered with the M13 tails will allow the use of the universal M13 forward and M13 reverse primers for sequencing, the sequence immediately after the M13 primer will be the target primer sequence, and so the SNP can be close to the 3' end of the target primer sequence.

How do I find primers for a specific exon using the Primer Designer Tool?

Exon counting is not standardized. Exon 1 often means the first exon on the transcript, but could also mean the first exon on the genome regardless of the sense of transcription. Within the Primer Designer Tool, the mouse over pop‐up‐box on the genomic viewer indicates the number of an exon. However, we recommend user confirmation to ensure that the exon number indicated is concordant with the exon counting system the user expects.

When using the Primer Designer Tool, what are the supported search terms for finding predesigned primers?

Within the Primer Designer Tool, the following search terms can be used:
- Assay ID: e.g., "Hs00326060_CE"
- SNP ID: e.g., "rs12096805"
- Entrez Gene ID: e.g., "6262"
- Gene Symbol: e.g., "JAK2"
- Gene Name: e.g., "Janus kinase 2"
- RefSeq Accession #: e.g., "NM_007294"
- GenBank mRNA: e.g., "CR536579.1"
- Protein ID: e.g,. "CAA66975.1"
- Cosmic ID: e.g., "121007"
The Primer Designer Tool also allows one to enter a sequence and mapping primer pairs will be returned.

How do I determine which area is sequenced by the predesigned primers ordered through the Primer Designer Tool?

Determining the area sequenced by the predesigned primers ordered via the Primer Designer Tool can be done in the following way:
For each amplicon, the amplicon coordinates are displayed. Clicking on "View Primer on Map" will display the location of the amplicons on the genome and in the context of transcripts.

What is the delivery time frame for predesigned primer pairs ordered via the Primer Designer Tool?

The typical delivery time frames for predesigned primer pairs ordered using the Primer Designer Tool for USA and Canada are as follows:
- Desalted primers: delivery within 3 business days
- HPLC-purified primers: delivery within 8 business days

- The typical delivery time frames for predesigned primer pairs ordered using the Primer Designer Tool for Europe, the Middle East, and Africa are as follows:
- Desalted primers: delivery within 3 business days
- HPLC-purified primers: delivery within 3-5 business days

Are your Ion Reporter Software annotations based on GRCh Build 38 or hg19?

If your AmpliSeq design has been created using hg38 as a reference, then you can create an ad-hoc workflow in Ion Reporter Software for analysis. All analysis and annotations will take into consideration hg38 as a reference. However, at this moment there are no hg38 workflows in Ion Reporte Software. The tools for analysis and annotations for hg19 will still be available.

Will there be a version of the Oncomine panels based on GRCh Build 38?

At this moment, there are no new versions of the Oncomine panels based on GRCh Build 38. The Oncomine Panels will still be based on hg19.

Will there be a new version of the ready to use AmpliSeq panels based on GRCh Build 38?

At this moment, there are no new versions of the ready to use AmpliSeq panels based on GRCh Build 38. The off-the-shelf panels will still be based on hg19.

Can I copy amplicons from a hg19 design to a GRCh Build 38 design (or vice versa)?

No, you cannot copy amplicons from an hg19 design to an hg38 design (or vice versa). Amplicons from a custom design can only be copied to another custom design associated with the same reference. It is not possible to copy amplicons to a custom design associated with a different reference.

Can I still annotate my old variants (based on hg19) with Ion Reporter Software?

Yes, the variant calling workflow based on hg19 will still be available in Ion Reporter Software. If your design was created using GRCh Build 38, you can also call and annotate variants using Ion Reporter Software.

Can I still design, order, and analyze old Ion AmpliSeq panels based on hg19?

Yes. The pipelines and tools for using hg19 as reference for design and analysis will still be available.

Will the Variant Analysis and Next-Generation Confirmation cloud apps support analysis of data using GRCh Build 38 primers?

Yes, the Variant Analysis and Next-Generation Confirmation cloud apps will support all primers that map to GRCh Build 38. You will be informed with an error message if the primer pair does not map to this release.

How can I analyze my CE data with hg19 primers after the transition to GRCh Build 38?

After transition to the GRCh Build 38 build, you should use software and analysis pipelines that still support hg19 to analyze your CE data. CE desktop software, such as Variant Reporter and SeqScape software, will continue to support hg19.

You can also perform analysis using the Variant Analysis (VA) or Next-Generation Confirmation (NGC) cloud apps, but you will need to create a reference using an hg19 Genbank file. If the hg19 Genbank file has annotations, VA or NGC will report out variants based on your provided annotations. In NGC, you will also need to provide the NGS variant file (vcf file) which is generated using hg19 as a reference.

If I search by the same coordinates I used for hg19 on the hg38 build, will I get the same primers?

We cannot guarantee that you will get the same primers if you search by the same coordinates you used for hg19 on the hg38 build.

Can I analyze Ion AmpliSeq assays or panels that use the hg19 human reference with the new GRCh build 38 human genome reference?

No, the AmpliSeq assays and panels that were created using hg19 as a reference should be analyzed with the tools and analysis pipelines created for hg19.

Do you have a conversion tool that converts from hg19 coordinates to GRCh Build 38 coordinates?

At this moment, we do not offer any conversion tools to convert coordinates between hg19 and GRCG Build 38. There are publicly available tools to convert coordinates, such as UCSC's LiftOver tool which can be accessed here: http://genome.ucsc.edu/cgi-bin/hgLiftOver

Can I download the GRCh Build 38 files from NCBI and use them directly for my analysis of Ion sequencing data?

We strongly recommend that you download our version of the GRCh Build 38 from our website to use for your analysis of Ion sequencing data. This version is the one that is assumed in all of our software applications.

Which "alt_sequence" does your version of GRCh Build 38 consider?

Our version of GRCh Build 38 only considers the chr22_KI270879v1_alt. This alt chromosome contains gene GSTT1 that was part of chr22 in hg19.

How is the GRCh Build 38 reference used by Thermo Fisher Scientific software different from the references publicly available from places like UCSC or NCBI?

The differences between the GRCh Build 38 reference used by Thermo Fisher Scientific software and the references publicly available from places like UCSC or NCBI are listed below:
- Repeat and SNP locations are soft-masked into lower case letters, while the ambiguous IUPAC bases, duplicated centromeric arrays, and chrY PAR regions are hard masked into 'N's.
- The version used by our software is based on GRCh Build 38.p2 (http://www.ncbi.nlm.nih.gov/assembly/GCF_000001405.28)
- The GRCh Build 38 reference contains chr1-22, chrX, chrY, chrM, and chr22_KI270879v1_alt.
- Contig chr22_KI270879v1_alt is hard masked except for region 269814-279356 (1-based).
- Gene GSTT1 is located at chr22_KI270879v1_alt:270308-278486.

Why is Thermo Fisher Scientific making the change to GRCh Build 38?

We are moving to GRCh Build 38 because it is a more accurate representation of the human genome and is the most recent genome version being accepted by the human genome community. Since many of the annotation databases have moved their content to correspond with the GRCh Build 38 coordinates, this change allows us to link to these newer annotations.

How is GRCh Build 38 essentially different from hg19?

The GRCh Build 38 is essentially different from hg19 in that the hg19 build is a single representation of multiple genomes whereas the GRCh Build 38 provides alternate sequences ("alt_sequences") for some genomic regions for which their variability prevents adequate representation by one single reference. Other differences between the builds include changes in the centromere representation, the mitochondrial genome, updated sequences, and an "Analysis Set" (FASTA format sequences in a package convenient for use by various Next Generation Sequence read alignment pipelines).

Is hg38 (Human Genome Version 38) the same genome version as GRCh Build 38?

Yes, hg38 and GRCh Build 38 are the same version of the human genome. GRCh Build 38 stands for "Genome Reference Consortium Human Reference 38" and is the primary genome assembly in GenBank; hg38 is the ID used for GRCh Build 38 in the context of the UCSC Genome Browser.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.