Learn More
Invitrogen™ M13 phage coat protein Monoclonal Antibody (A4C11), HRP

Description
M13 phage coat protein Monoclonal Antibody for ELISA
Specifications
Specifications
| Antigen | M13 phage coat protein |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | A4C11 |
| Concentration | 2 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | Attachment protein G3P; G3P; Gene 3 protein; III; Minor coat protein |
| Host Species | Mouse |
| Immunogen | Synthetic peptide within bacteriophage M13 aa 1-50. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.