Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ M13 phage coat protein Monoclonal Antibody (A5B3), HRP
GREENER_CHOICE

Catalog No. PIMA536125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536125 Supplier Invitrogen™ Supplier No. MA536125
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

M13 phage coat protein Monoclonal Antibody for ELISA

M13 antibodies have numerous uses in screening phage display libraries, especially the screening of single chain antibody libraries. The display of repertoires of antibody fragments on the surface of filamentous phage offers a new way to produce immunoreagents with defined specificities. Phage derived antibody fragments offer a number of advantages over mouse monoclonal antibodies, such as better clearance from the blood, the possibility to select from human combinatorial libraries and the relative ease by which such fragments can be manipulated. The phage display technique thus facilitates the selection of antibody fragments of therapeutic value or research interest. Antibodies to M13 filamentous phage coat proteins are instrumental in the selection and detection of phages expressing specific antibody fragments or peptide sequences at their surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen M13 phage coat protein
Applications ELISA
Classification Monoclonal
Clone A5B3
Concentration 2 mg/mL
Conjugate HRP
Formulation PBS with 40% glycerol, 0.1% BSA and no preservative; pH 7.4
Gene Alias Attachment protein G3P; G3P; Gene 3 protein; III; Minor coat protein
Host Species Mouse
Immunogen Synthetic peptide within bacteriophage M13 aa 1-50.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.