Learn More
Invitrogen™ M13 phage coat protein g8p Monoclonal Antibody (RL-ph2)
Shop All Thermo Scientific Products

Description
MA1-06604 detects M13 in phage and phage infected cells. MA1-06604 has sucessfully been used in Western blotting, flow cytometry, ELISA, and immunocytochemistry procedures. The MA1-06604 immunogen isM13 phage coat protein g8p. Store at 4°C or in small aliquots at -20°C.
Specifications
Specifications
| Antigen | M13 phage coat protein g8p |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | RL-ph2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Host Species | Mouse |
| Immunogen | Isolated M13 phage coat proteins. |
| Purification Method | Protein G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.