Learn More
Applied Biosystems™ M13 Sequencing Reverse Primer, HPLC
Description
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.
Specifications
Specifications
| Delivery Type | Single Tube, Dried-down |
| For Use With (Equipment) | Mass Spectrometry |
| Description | M13 Reverse Sequencing Primer, 50 nmole, HPLC Purification, 900pmole Minimal Spec Yield, 5' CAGGAAACAGCTATGACC Sequence, Delivered in a Single tube, Dried-down, Used for BigDye™ Cycle Sequencing Primers |
| Product Type | M13 Reverse Sequencing Primer |
| Protein Degradation Sequence | 5' CAGGAAACAGCTATGACC |
| Primer | M13 Reverse Sequencing |
| Quantity | 50 nmol |
| Grade | HPLC |
| For Use With (Application) | BigDye™ Cycle Sequencing Primers |
| Form | Solid |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.