Learn More
Worldwide™ Protective Products Low-Lint Seamless Knit Polyester Gloves
Catalog No. 18701024 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-701-024 Medium
18-701-022 X-Large
18-701-026 X-Small
18-701-025 Small
18-701-023 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 18-701-024 Supplier Worldwide™ Protective Products Supplier No. M13PXYLBM
  • $64.95
    $62.65 / Dozen of 12
    Save $2.30
    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $5,013.43
    $4,835.65 / Case of 30 DZ
    Save $177.78
    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Gloves are ideal for protecting products from lint or other contaminants. Design is light enough to be worn as a liner, and provides high dexterity and fingertip sensitivity for precise hand work.
  • Gloves deliver the high dexterity and fingertip sensitivity required for paint rooms, QC, and inspection work
  • Can be worn under metal mesh, leather or disposable gloves as a liner
  • Made from 100% filament, 13-gauge polyester
  • Protects products from lint, dirt, and other contaminants
  • Lightweight, conforming design maximizes wearer comfort
  • Pre-laundered and bagged to maintain product integrity
  • Color-coded hems for easy size identification
Specifications
Construction Seamless Knit
Coating Material Uncoated
Color White
Disposable Limited-use
Hand Specific
Latex Free Yes
Material Polyester
Product Type Inspection Gloves
Size Medium
Sterility Non-sterile
Glove Cuff Style Continuous Knit
Series M13PXY-LB
Yarn Gauge 13
Length (English) 9.5 in.
Length (Metric) 24.1 cm