Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Methylhexanamine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco M1571ML
Description
DMAA, also known as dimethylamylamine, is a stimulant sold as a dietary supplement under trade names including Geranamine and Floradrene. Since late 2009 DMAA has emerged as a club/designer drug in party pills and bath salt blends. Many professional and amateur sports bodies, such as the World Anti-Doping Agency (WADA), have banned methylhexanamine as a performance-enhancing substance. This certified solution standard is suitable for a variety of testing applications, from clinical toxicology, urine drug testing, and forensic analysis to sports testing and analysis of DMAA-based dietary supplements.
Specifications
67-56-1, 13803-74-2 | |
MFCD09038354 | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
9.7°C (49.5°F) |
C7H17N · HCl | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
151.68 | |
1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : -20°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?Share a Content Correction