Learn More
ILEX LIFE SCIENCES LLC M199 EBSS P04-07050 500 ML
Supplier: ILEX LIFE SCIENCES LLC P0407050
PAN-Biotech Medium 199 (M199) w: EBSS, w: L-Glutamine, w: 2.2 g/L NaHCO3 (500 ml). EBSS: Contains Earle′s Balanced Salt Solution (EBSS). BICARBONATE: With 2.2 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With L-glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Medium 199, also known as M199, was originally developed to assay the nutrient demand of embryonic chicken fibroblasts. It also works very well with cells from many different animal species. Serum should be added for long term cultures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.