Learn More
Simport™ Scientific M480 Disposable Embedding Cassettes in Quickload™ Stacks
Catalog No. 22038036 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Green
Lilac
Orange
Peach
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-036 Green
22-038-037 Yellow
22-038-039 Peach
22-038-042 Lilac
22-038-043 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-038-036 Supplier Simport™ Scientific Supplier No. M4804T
Add to Cart
Add to Cart
This item is not returnable. View return policy
M480 Disposable Embedding Cassettes in Quickload™ Stacks for holding and identifying tissue samples in processing/embedding procedures.
  • In Quickload™ stacks
  • 45° Anterior writing area
  • Accept standard metal lids (cat# M481)
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Quantity 2000/Pk.
Color Green
Material Acetal