Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ M4K2163 dihydrochloride
GREENER_CHOICE

Catalog No. p-7250158
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-541-0 10 mg
77-545-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 77-541-0 Supplier Tocris Bioscience™ Supplier No. 7754/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective ALK2 inhibitor

M4K2163 is a potent and selective activin receptor-like kinase 2 (ALK2) inhibitor (IC50 values are 19 nM in vivo and 98 nM in vitro). Orally bioavailable and brain penetrant.

Specifications

Chemical Name or Material 1-[4-[5-(4-Fluoro-3,5-dimethoxyphenyl)-4-methyl-3-pyridinyl]phenyl]piperazine dihydrochloride
Quantity 10 mg
Target ALK/CD246
Molecular Formula C24H26FN3O20.2 HCl
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.