Learn More
CPC Scientific H-Cys-Asp-Ala-Thr-Cys-Gln-Phe-Arg-Lys-Ala-Ile-Asp-Asp-Cys-Gln-Lys-Gln-Ala-His-His-Ser-Asn-Val-Pro-Gly-Asn-Ser-Val-Phe-Lys-Glu-Cys-Met-Lys-Gln-Lys-Lys-Glu-Phe-Lys-Ala-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PACR001A
Encompass
PAC1 receptor antagonistONE-LETTER SEQUENCE: CDATCQFRKAIDDCQKQAHHSNVPGNSVFKECMKQKKEFKA-NH2 (Cys1 & Cys5, Cys14 & Cys32 bridge)MOLECULAR FORMULA: C199H314N62O60S5MOLECULAR WEIGHT:4695.39STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: O.Moro et al., J. Biol. Chem., 274, 23103 (1999); E.A.Lerner et al., Peptides, 28, 1651 (2007)
Catalog No. 50-211-0068
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.