Learn More
Epigentek Inc N6-methyladenosine (m6A) Polyclonal Antibody | 20uL
SDP
Supplier:  Epigentek Inc A1801020
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Chemical compounds corresponding to N6-methyladenosine / m6A.. Tested Applications: ELISA, DB, IF, IP, MeRIP. Reactivity: Human, Mouse, Broad Range. Alternative Names: Anti-N6-methyladenosine, N6-methyladenosine, N6(methylnitroso)adenosine, m6A, N6-methyladenosine, 6mA.
Catalog No. NC1628732
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.