Learn More
Abcam m6A RNA Methylation Assay Kit, Fluorometric, 48 Tests
Description
- Assay time: 3 hr 45 min
- Sensitivity: 5 pg/well
- Detection method: Fluorescent
- Results type: Quantitative
- Sample types: Tissue, suspension cells, adherent cells, and other biological fluids
- Usage: Compatible with microplate reader
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | N6-methyladenosine (RNA) |
| Run Time | 3 hr. 45 min. |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 48 Tests |
| Detection Location | In-Solution Detection |
| Detection Method | Fluorescence |
| Sample Type | Tissue, Suspension Cells, Other biological fluids, Adherent Cells |
| Quantity | 48 Tests |
| For Use With (Application) | m6A RNA methylation measurement |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.