Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam m6A RNA Methylation Assay Kit, Fluorometric, Quantitative, 1x96 Tests
Description
- Detection Method: Fluorescent
- Assay Time: 3 hr 45 min
- Sensitivity: 5 pg/well
- Results Type: Quantitative
- Sample Types: Tissue, suspension cells, adherent cells, and other biological fluids
- Usage: Suitable for use with a microplate reader
- Citations: Cited in 4 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | N6-methyladenosine (RNA) |
| Run Time | 3 hr. 45 min. |
| No. of Reactions | 96 Tests |
| For Use With (Equipment) | Microplate Reader |
| Detection Location | In-Solution Detection |
| Format | Kit |
| Detection Method | Fluorescence |
| Sample Type | Tissue, Suspension Cells, Other Biological Fluids, Adherent Cells |
| Quantity | 96 Tests |
| For Use With (Application) | Measure N6-methyladenosine (m6A) in cell/tissue extracts, cell samples and biofluids |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?