Learn More
Teknova M9 Minimal Salt Broth with 2% Casamino Acids,10% Glycerol, and 0.5µg/mL Thiamine. 1000mL, Sterile 2 pack.
SDP
Supplier:  Teknova M82452PK
Encompass_Preferred
1X M9 Salts, 2.0% Casamino Acids, 10.0% Glycerol, 0.5µg/mL Thiamine, 0.2mM Magnesium Sulfate, 0.1mM Calcium Chloride
Catalog No. 50-238-7660
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.