Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BDP Recombinant Rabbit Monoclonal Antibody (008)
GREENER_CHOICE

Catalog No. pima535969
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535969 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535969 Supplier Invitrogen™ Supplier No. MA535969
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

BDP Recombinant Monoclonal Antibody for Western Blot

This gene encodes a member of the ARID family of DNA-binding proteins. The encoded protein is homologous with two proteins that bind to the retinoblastoma gene product, and also with the mouse Bright and Drosophila dead ringer proteins. A pseudogene on chromosome 1p31 exists for this gene. Members of the ARID family have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BDP
Applications Western Blot
Classification Recombinant Monoclonal
Clone 008
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ARID3B
Gene Accession No. Q8IVW6
Gene Alias ARID domain-containing protein 3B; ARID3B; AT rich interactive domain 3B (Bright like); AT rich interactive domain 3B (BRIGHT- like); AT rich interactive domain 3B (BRIGHT-like); AT-rich interaction domain 3B; AT-rich interactive domain-containing protein 3B; Bdp; bright and dead ringer gene product homologous protein Bdp; bright and dead ringer protein; Bright-like; Bright-like protein; dead ringer homolog 2; Dri2; DRIL2
Gene Symbols ARID3B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human ARID3B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10620
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.