Learn More
Bioworld Maackia amurensis Lectin (MAA/MAL I) - FITC
Supplier:  Bioworld 217610361
Encompass
Affinity purified Maackia amurensis lectin (MAA) is a tetramer formed by MAA1 or MAL (a 37kD subunit called Maackia amurensis leukoagglutinin) and MAA2 or MAH (a 32 kD subunit called Maackia amurensis hemagglutinin).Technical Specifications:Ligand Source:Maackia amurensisMolecular Weight: 130 kDaCarbohydrate Specificity:Siaα1,3Galβ4GalNAcInhibiting/Eluting Sugar:LactoseCarbohydrate for Elution: 0.2MLactoseBlood Group Specificity: N/AConjugate: FITCForm: LiquidIsoelectric (pI): 4.7Storage: -20°CApplication note: N/A
Catalog No. 50-199-151
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.