Learn More
Bioworld Maackia amurensis Lectin (MAA/MAL I) - Rhodamine (TRITC)
Supplier: Bioworld 217610371
Technical Specifications:Ligand Source:Maackia amurensisMolecular Weight:130 kDaCarbohydrate Specificity:Siaα1,3Galβ4GalNAcInhibiting/Eluting Sugar:LactoseCarbohydrate for Elution:0.2MLactoseBlood Group Specificity:N/AConjugate: RhodamineForm:LiquidIsoelectric (pI):4.7Storage: -20°CApplication note: N/A
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.