Learn More
Bioworld MAACKIA AMURENSIS LECTINMAA)
Supplier:  Bioworld 215100071
Encompass_Preferred
Maackia amurensis lectin (MAA) Affinity purified Maackia amurensis lectin is a tetramer formed by a 37 kD subunit called MAL (Maackia amurensis leukoagglutinin) also known as MAA1 and a 32 kD subunit called MAH ((Maackia amurensis hema...
Catalog No. 50-198-739
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.