Learn More
Bio X Cell InVivoMAb anti-human NKG2D (CD314), Clone: 1D11, Size: 1 mg
Supplier:  Bio X Cell BE0351R001MG
Encompass_Preferred
InVivoMAb anti-human NKG2D (CD314), Clone: 1D11, Size: 1 mg, Isotype: Mouse IgG1, κ, Applications: in vitro NKG2D blocking, Immunoprecipitation, Flow cytometry
Catalog No. 50-217-2863
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.