Learn More
Bio X Cell InVivoMAb anti-mouse CD16/CD32, Clone: 2.4G2, Size: 25 mg
Supplier:  Bio X Cell BE0307R025MG
Encompass_Preferred
InVivoMAb anti-mouse CD16/CD32, Clone: 2.4G2, Size: 25 mg, Isotype: Rat IgG2b, κ, Applications: in vitro Fc receptor blocking, in vivo Fc receptor blocking
Catalog No. 50-152-8494
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.