Learn More
Bio X Cell InVivoMab anti-human/monkey CD20, Clone: 2H7, Size: 1 mg
Supplier:  Bio X Cell BE0276R001MG
Encompass_Preferred
InVivoMab anti-human/monkey CD20, Clone: 2H7, Size: 1 mg, Isotype: mouse IgG2b, κ, Applications: in vivo B cell depletion in hCD20 Tg mice, Immunohistochemistry (frozen), Immunoprecipitation, Flow cytometry
Catalog No. 50-152-8456