Learn More
Bio X Cell InVivoMab anti-mouse CD28, Clone: PV-1, Size: 1 mg
Supplier:  Bio X Cell BE00155R001MG
Encompass_Preferred
InVivoMab anti-mouse CD28, Clone: PV-1, Size: 1 mg, Isotype: Armenian hamster IgG, κ, Applications: in vitro T cell stimulation/activation
Catalog No. 50-152-8242
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.