Learn More
Axxora LLC MAB TO CD39 (HUMAN) 100UG
SDP
Supplier:  Axxora LLC ANC188020
Encompass
Monoclonal Antibody to CD39, Clone: BU61, Species Reactivity: Human, Specifity: Recognizes human CD39, Isotype: Mouse IgG1, Form: Liquid, Immunogen: Human WM-1cell line, Application: FC, IHC, IP, WB, 100ug
Catalog No. AANC188020
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.