Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MabCaptureC™ High Capacity Protein A Resin

Catalog No. A40004566
Change view
Click to view available options
Quantity:
100 mL
2 Plates
25 mL
5 Columns
5 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40004566 5 Columns
PIA53031 5 mL
PIA53033 25 mL
PIA53034 100 mL
A40004564 2 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A40004566 Supplier Thermo Scientific™ Supplier No. A40004566
Add to Cart
Edge
Add to Cart

MabCaptureC High-Capacity Protein A Spin Columns provide a fast, ready-to-use, method for capturing monoclonal antibodies (mAbs) and recombinant Fc-tagged fusion proteins.

GoPure MabCaptureC Screening Plates, 96-well, 50 μL, contain MabCaptureC High-Capacity Protein A Resin, a high-performance chromatography resin for capturing monoclonal antibodies (mAbs) and recombinant Fc-tagged fusion proteins. It provides high specific-binding capacity and enhanced alkaline-stability characteristics under clean-in-place conditions (CIP), for more robust purification and resin regeneration. These ready-to-use, 96-well spin plates offer a high-throughput format for rapid antibody purification, screening, and evaluating multiple conditions in a single plate experiment.

Features of MabCaptureC High Capacity Protein A Resin

  • High binding capacity—60 mg hIgG/mL at 5-minite residence (80 mg/mL at 10-minute residence)
  • Alkaline stability—engineered to withstand common CIP procedures with 0.2 M NaOH, enabling efficient cleaning of the chromatography resin and improving product quality
  • Animal component-free raw materials
  • Versatility—clean antibody or Fc-fusion protein preparations from cell culture supernatant, serum, or ascites
  • Stability—high-affinity Protein A is securely immobilized to high purity agarose to prevent leaching of ligand, target antibody, or base support
  • Flexible— available in multiple formats, including bulk loose resin, spin columns, and 96-well spin plates

The Protein A ligand, which is produced in yeast, is immobilized on a rigid and uniformly sized agarose bead with an average particle size of 75 μm. The uniformity of the particle size, in turn, delivers superior performance characteristics over traditional resins.

MabCaptureC High-Capacity Protein A Resin, alkaline-stable, is suitable for the purification of mAbs as well as a subpopulation of scFv's and Fab fragments that contain a VH3 domain.

Available formats

  • Resin slurries—10 mL, 50 mL, and 200 mL of 50% slurry
  • Spin columns—10 mL spin columns containing 1 mL of settled resin, supplied at 5 mL 20% slurry
  • GoPure 96-well spin plates—96-well spin plates with 20 μL or 50 μL of settled resin
TRUSTED_SUSTAINABILITY

Specifications

Product Type Spin Column
Column Type Agarose Resin, Affinity
Format Spin Column
Stationary Phase Protein A
Quantity 5 Columns
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.