Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva MabSelect PrismA Chromatography Resin

Catalog No. 09928073 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-928-073 25 mL
09-928-074 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-928-073 Supplier Cytiva Supplier No. 17549801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protein A Chromatography Resin

MabSelect PrismA has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing future demands in mAb processing to be met.

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 80mg human IgG/mL resin at 6 min residence time. 65 mg human IgG/mL resin at 4 min residence time.
Matrix Highly cross-linked agarose, spherical.
Content And Storage 2 to 8°C, 20% ethanol
Max. Operating Pressure 300 cm/hr.
Particle Size 60 μm
Quantity 25 mL
For Use With (Application) Antibody affinity chromatography
Type Chromatography Resin
Ligand Type Alkaline-stabilized protein A-derived (E.coli).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.