Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva MabSelect™ VL Protein L Resin

Catalog No. 09928201 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-928-201 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-928-201 Supplier Cytiva Supplier No. 17542002
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Protein L resin for affinity capture of antibody Fabs, bispecifics, and other variants containing a kappa light chain.

MabSelect™ VL resin with a redesigned protein L ligand for high binding capacity and improved alkaline stability. For affinity capture of antibody Fabs, bispecifics, and other variants containing a kappa light chain.

  • High dynamic binding capacity protein L resin for efficient capture of antibody Fabs, bispecifics, and variants containing a kappa light chain.
  • Stable when cleaned with 0.1 M NaOH, reducing risk for bioburden incidents.
  • Provides good resolution for product-related impurities in the capture of bispecific antibodies.
  • Can withstand cleaning with 0.1 M NaOH
  • Available in 1 mL and 5 mL HiTrap™ prepacked columns
TRUSTED_SUSTAINABILITY

Specifications

Product Type Resin
Product Line HiTrap
Column Type Agarose Resin, Affinity
Format Prepacked Column
Stationary Phase Protein L
Binding Capacity 70 mg
Ligand Alkaline stabilized protein L-derived (E. coli)
Matrix Spherical Highly Cross-linked Agarose
Particle Size 60 μm
Content And Storage 2°C to 8°C
Quantity 200 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.