Learn More
Axxora LLC MABTOFAS(H)(NONINDUC) 120TESTS
SDP
Supplier:  Axxora LLC ANC316040
Encompass
Monoclonal Antibody to Fas (human) (ANC95.1/5E2),120 tests, Synonyms anti-CD95 (human) MAb (ANC95.1/5E2), anti-APO-1 (human) MAb (ANC95.1/5E2), Isotype: Mouse IgG1, Formulation: Liquid. Purified antibody contains 0.5mg/ml gentamycin sulfate
Catalog No. AANC316040
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.