Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MacConkey Agar

Catalog No. 091006017 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
09-100-6017 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-100-6017 Supplier MP Biomedicals™ Supplier No. 091006017
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MacConkey Agar is powder media used for isolating enteric bacilli.

  • Medium used for the isolation of enteric bacilli based on the fermentation of lactose.
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Media Type MacConkey Agar
Quantity 500 g
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.