Learn More
Teknova MacConkey Agar Plates. 150mm Plates, Sterile. 4 pack.
SDP
Supplier:  Teknova M01504PK
Encompass_Preferred
1.70% Tryptone, 0.30% Peptone,1.00% Lactose, 0.15% Bile Salts #3,0.50% Sodium Chloride, 1.35% Agar,30µg/mL Neutral Red, 1µg/mL Crystal Violet
Catalog No. 50-238-7826
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.