Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MACF1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7142967
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8055 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8055 Supplier Bethyl Laboratories, Inc Supplier No. A304564AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Isoform 2 is a F-actin-binding protein which may play a role in cross-linking actin to other cytoskeletal proteins and also binds to microtubules. Plays an important role in ERBB2-dependent stabilization of microtubules at the cell cortex. Acts as a positive regulator of Wnt receptor signaling pathway and is involved in the translocation of AXIN1 and its associated complex (composed of APC, CTNNB1 and GSK3B) from the cytoplasm to the cell membrane. Has actin-regulated ATPase activity and is essential for controlling focal adhesions (FAs) assembly and dynamics. May play role in delivery of transport vesicles containing GPI-linked proteins from the trans-Golgi network through its interaction with GOLGA4. Plays a key role in wound healing and epidermal cell migration. Required for efficient upward migration of bulge cells in response to wounding and this function is primarily rooted in its ability to coordinate MT dynamics and polarize hair follicle stem cells.

Specifications

Antigen MACF1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MACF1
Gene Accession No. Q9QXZ0, Q9UPN3
Gene Alias 620 kDa actin binding protein, 620 kDa actin-binding protein, ABP620, ACF7, actin cross-linking factor, actin cross-linking family protein 7, FLJ45612, FLJ46776, KIAA0465, KIAA1251, MACF, macrophin 1, macrophin 1 isoform, Macrophin-1, OFC4, trabeculin-alpha
Gene Symbols MACF1
Host Species Rabbit
Immunogen Between 1975 and 2025
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11426, 23499
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.