Learn More
Ancare Corporation MACROLON BTTL 32OZ SCRW 3X3X9H
SDP
Supplier:  Ancare Corporation MST32
Encompass
Material: autoclave polycarbonate or polysulfone.Accepts Ancare PCST screw tops, number 6.5 stopper or double seal stopper /S-100/S-101/.Graduated in ounces and milliliters.
Catalog No. NC2709452