Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophage Mouse anti-Human, Unconjugated, Clone: HAM-56, MilliporeSigma™

Numéro de catalogue. MABF22521MI missing translation for 'brandsLinkText'
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MABF22521MI 100 μg
MABF22522MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MABF22521MI Fournisseur MilliporeSigma Code fournisseur MABF2252100UG
Il en reste null
ajouter au panier
ajouter au panier

Mouse Monoclonal Antibody

Macrophage Monoclonal antibody specifically detects Macrophage in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Spécifications

Antigen Macrophage
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HAM-56
Conjugate Unconjugated
Gene Alias Alveolar macrophages
Host Species Mouse
Immunogen Human alveolar macrophages.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone HAM-56 is a mouse monoclonal antibody that specifically detects human macrophages.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgM κ
Afficher plus de résultats Afficher moins de résultats
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.