Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophage Mouse anti-Human, Unconjugated, Clone: HAM-56, MilliporeSigma™

Catalog No. MABF22522MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22522MI 25 μg
MABF22521MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22522MI Supplier MilliporeSigma Supplier No. MABF225225UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Macrophage Monoclonal antibody specifically detects Macrophage in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Macrophage
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HAM-56
Conjugate Unconjugated
Gene Alias Alveolar macrophages
Host Species Mouse
Immunogen Human alveolar macrophages.
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone HAM-56 is a mouse monoclonal antibody that specifically detects human macrophages.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.