Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MadMax™ II Synthetic Leather Palm Mechanics Gloves with TPR Padding

Catalog No. 19824159 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-824-159 2X-Large
19-824-155 Small
19-824-156 Medium
19-824-157 Large
19-824-158 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-824-159 Supplier PIP™ Supplier No. 1204050/XXL
This item is not returnable. View return policy

Palms are constructed of synthetic leather with PVC dots for enhanced grip capabilities in dry, or light oil applications.

  • Synthetic leather palm provides dexterity, and enhances grip in dry or light oil applications
  • PVC dot grip on palm and fingertips for extra cut/abrasion resistance
  • Thermoplastic Rubber (TPR) molded finger, thumb, and back hand guards for maximum protection
  • Reinforced thumb crotch for extended glove life, enhanced comfort, and increased abrasion resistance
  • Airprene wrist is made with breathable neoprene for comfort
  • Synthetic leather upper and heavy lycra fabric bottom fourchettes
  • Foam padded knuckle protection

Specifications

Back Material TPR Padding
Cuff Material Airprene
Coating Material PVC Dots (1 Side)
Color Hi-vis Yellow
Hand Specific
Material Synthetic Leather, Spandex, Lycra
Product Type Mechanic's Gloves
Size 2X-Large
For Use With (Application) Construction, Auto Repair, Manufacturing, Mining, and Landscaping
Glove Cuff Color Black
Glove Cuff Style Open
Series 120-4050
Length (English) 10.8 in.
Length (Metric) 27.3 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.