Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Mad Max™ Thermo Waterproof Synthetic Leather Gloves with TPR Padding

Catalog No. 19824163 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Large
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-824-163 2X-Large
19-824-162 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-824-163 Supplier PIP™ Supplier No. 1204070/XXL
This item is not returnable. View return policy

Gloves are lined with 3M™ Thinsulate™ for thermal protection, without the bulkiness of fleece. The synthetic leather palm enhances grip in in dry, or light oil applications.

  • Synthetic leather palm provides dexterity, and enhances grip
  • PVC sandy grip on palm and fingertips provides additional cut/abrasion resistance
  • 3M™ Thinsulate™ lining provides thermal protection without the bulk of fleece
  • HIPORA™ liner is waterproof, windproof, and breathable
  • Thermoplastic Rubber (TPR) molded finger, thumb, and back hand guards for maximum protection
  • Reinforced thumb crotch for extended glove life, enhanced comfort, and increased abrasion resistance
  • Airprene wrist is made with breathable neoprene for comfort
  • Synthetic leather upper, and heavy Lycra™ fabric bottom fourchettes
  • Hi-vis coloring and reflective back

Specifications

Cuff Material Nylon Airprene
Color Hi-vis Green, Black
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size 2X-Large
For Use With (Application) Construction, Auto Repair, Manufacturing, and Landscaping
Glove Cuff Color Black
Glove Cuff Style Open
Liner Material 3M™ Thinsulate™
Lining HIPORA™
Series 120-4070
Length (English) 11.8 in.
Length (Metric) 30 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.