Learn More
Invitrogen™ MAD1 Monoclonal Antibody (9B10)

Description
MAD1 Monoclonal Antibody for ICC/IF, Flow, IP
Specifications
Specifications
| Antigen | MAD1 |
| Applications | Flow Cytometry, Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 9B10 |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | MAD1L1 |
| Gene Accession No. | Q9Y6D9 |
| Gene Alias | AI173502; AW550425; coiled-coil domain-containing protein MAD1; hMAD1; HsMAD1; Mad1; MAD1 mitotic arrest deficient 1-like 1; MAD1 mitotic arrest deficient like 1; MAD1 mitotic arrest deficient-like 1; MAD1 mitotic arrest deficient-like 1 (yeast); MAD1L1; MAD1-like protein 1; mitotic arrest deficient 1-like 1; mitotic arrest deficient 1-like protein 1; Mitotic arrest deficient protein 1; Mitotic checkpoint MAD1 protein homolog; mitotic spindle assembly checkpoint protein MAD1; mitotic-arrest deficient 1, yeast, homolog-like 1; MXD1; PIG9; Spindle assembly checkpoint component MAD1; Tax-binding protein 181; TP53I9; tumor protein p53 inducible protein 9; TXBP181; YGL086W |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.