Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588148 Supplier Invitrogen™ Supplier No. PA588148
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGLEKARQQL QEELRQVSGQ LLEERKKRET HEALARRLQK RVLLLTKERD GMRAILGSYD SELTPAEYSP QLTRRMREAE DMVQKVHSHS AEMEAQLSQA LEELGGQKQR ADMLEMELKM LKSQSSSAEQ SFLFSREEAD TLRLKVEELE GERSRLEEEK RMLEAQLERR ALQGDYDQSR TKVLHMSLNP TSVARQRLRE DHSQLQAECE RLRGLLRAME RGGTVPADLE AAAASLPSSK EVAELKKQVE SAELKNQRLK EVFQTKIQEF RKACYTLTGY QIDITTENQY RLTSLYAEHP GDCLIFKATS PSGSKMQLLE TEFSHTVGEL IEVHLRRQDS I; Positive Samples: HeLa, HepG2, MCF7; Cellular Location: Chromosome, Cytoplasm, Nucleus, centromere, centrosome, cytoskeleton, kinetochore, microtubule organizing center, spindle.

MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAD1L1
Gene Accession No. Q9Y6D9
Gene Alias AI173502; AW550425; coiled-coil domain-containing protein MAD1; hMAD1; HsMAD1; Mad1; MAD1 mitotic arrest deficient 1-like 1; MAD1 mitotic arrest deficient like 1; MAD1 mitotic arrest deficient-like 1; MAD1 mitotic arrest deficient-like 1 (yeast); MAD1L1; MAD1-like protein 1; mitotic arrest deficient 1-like 1; mitotic arrest deficient 1-like protein 1; Mitotic arrest deficient protein 1; Mitotic checkpoint MAD1 protein homolog; mitotic spindle assembly checkpoint protein MAD1; mitotic-arrest deficient 1, yeast, homolog-like 1; MXD1; PIG9; Spindle assembly checkpoint component MAD1; Tax-binding protein 181; TP53I9; tumor protein p53 inducible protein 9; TXBP181; YGL086W
Gene Symbols MAD1L1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 360-700 of human MAD1/MAD1/MAD1L1 (NP_0010138591).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8379
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.