Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAD2L1BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531097
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531097 Supplier Invitrogen™ Supplier No. PA531097
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HepG2, HCT116. Predicted reactivity: Rat (82%), Rhesus Monkey (95%), Chimpanzee (99%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene was identified as a binding protein of the MAD2 mitotic arrest deficient-like 1 (MAD2/MAD2L1). MAD2 is a key component of the spindle checkpoint that delays the onset of anaphase until all the kinetochores are attached to the spindle. This protein may interact with the spindle checkpoint and coordinate cell cycle events in late mitosis. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD2L1BP
Applications Western Blot
Classification Polyclonal
Concentration 0.79 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene MAD2L1BP
Gene Accession No. Q15013
Gene Alias 0610009D16Rik; 2510031P20Rik; AU018945; AW550977; C76045; caught by MAD2 protein; CMT2; CMT2A; CMT2B; KIAA0110; MAD2L1 binding protein; MAD2L1-binding protein; Mad2l1bp; Mad2lbp; mKIAA0110; p31(comet); RP1-261G23.6
Gene Symbols MAD2L1BP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MAD2L1BP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9587
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.