Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAD2L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549352
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549352 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549352 Supplier Invitrogen™ Supplier No. PA549352
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAD2L2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. The encoded protein, which is similar to MAD2L1, is capable of interacting with ADAM9, ADAM15, REV1, and REV3 proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD2L2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MAD2L2
Gene Accession No. Q9D752, Q9UI95
Gene Alias 2310033C13Rik; G1-453-4; hREV7; MAD2 (mitotic arrest deficient, yeast, homolog)-like 2; MAD2 mitotic arrest deficient-like 2; MAD2 mitotic arrest deficient-like 2 (yeast); Mad2b; Mad2l2; MAD2-like protein 2; Mitotic arrest deficient 2-like protein 2; mitotic arrest deficient homolog-like 2; mitotic spindle assembly checkpoint protein MAD2B; polymerase (DNA-directed), zeta 2, accessory subunit; POLZ2; repro22; REV7; REV7 homolog; RP3-330O12.4
Gene Symbols MAD2L2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 198-231 amino acids from the C-terminal region of human MAD2L2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10459, 71890
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.