Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAD3 Recombinant Rabbit Monoclonal Antibody (JE48-14)
GREENER_CHOICE

Catalog No. PIMA536137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536137 Supplier Invitrogen™ Supplier No. MA536137
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

MAD3 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the Myc superfamily of basic helix-loop-helix leucine zipper transcriptional regulators. The encoded protein forms a heterodimer with the cofactor MAX which binds specific E-box DNA motifs in the promoters of target genes and regulates their transcription. Disruption of the MAX-MXD3 complex is associated with uncontrolled cell proliferation and tumorigenesis. Transcript variants of this gene encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE48-14
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MXD3
Gene Accession No. Q62912, Q80US8, Q9BW11
Gene Alias 4631412E13Rik; bHLHc13; class C basic helix-loop-helix protein 13; FLJ35523; MAD3; max dimerization protein 3; Max dimerizer 3; Max-associated protein 3; Max-interacting transcriptional repressor MAD3; MGC2383; Mxd3; Myx
Gene Symbols MXD3
Host Species Rabbit
Immunogen Recombinant protein within Human MAD3 aa 76-206.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17121, 252915, 83463
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.