Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MADD Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7144505
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2420 10 μL
50-157-2419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2420 Supplier Bethyl Laboratories, Inc Supplier No. IHC00621T
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine, Dog, Horse, Guinea pig_10141, Panda, Orangutan, Monkey, Gorilla and Ch

MAP kinase-activating death domain protein (MADD) was initially identified as the type 1 tumor necrosis factor receptor (TNFR1) associated protein though their death domains. Overexpression of MADD activates MAP kinases ERK and JNK and induces the phosphorylation of cytosolic phospholipase A2. MADD shares 98% identity with DENN (for differentially expressed in neoplastic vs. normal cells), which was recently identified as a substrate for c-jun N-terminal kinase 3 (JNK3). MADD has greater than 94% overall identity to a GDP/GTP exchange protein Rab3-GEP. MADD is 87% identical to KIAA0358, a brain protein of unknown function. Identification of MADD as a component of the TNFR1 signaling complex and the similarity between MADD and Rab3-GEP provides a connection between TNFR1 activation and downstream MAP kinase activity through a guanine-nucleotide exchange protein.

Specifications

Antigen MADD
Applications Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MADD
Gene Accession No. Q8WXG6
Gene Alias MAP-Kinase Activating Death Domain, DENN, Differentially Expressed in Neoplasmic vs. Normal Cells
Gene Symbols MADD
Host Species Rabbit
Immunogen Between 1597 and 1647
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8567
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.