Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MADD Recombinant Rabbit Monoclonal Antibody (JE63-62)
GREENER_CHOICE

Catalog No. PIMA544786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544786 Supplier Invitrogen™ Supplier No. MA544786
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MADD Recombinant Monoclonal Antibody for Western Blot

Tumor necrosis factor receptor 1 (TNFR1) contains a death receptor domain that mediates interactions involved in downstream signaling of TNF. A novel protein, MADD (mitogen activated protein (MAP)-kinase activating death domain protein) has recently been shown to associate with the death domain of TNFR1 through its own C-terminal death domain. This interaction is critical for TNFR1 signal generation. Overexpression of MADD activates the MAP (Mitogen Activated Protein) kinases extracellular signal-related kinase (ERK) and c-Jun N-terminal kinase (JNK) and induces the phosphorylation of cytosolic phospholipase A2. This discovery suggests that MADD links tumor necrosis factor TNF and MAP kinase activation and arachidonic acid release. MADD has a 98% homology with another protein, differentially expressed in neoplasmic vs. normal cells (DENN). DENN is a substrate for JNK3, and is important in the hypoxia and stress induced intracellular signalling pathways. MADD also has a 93% sequence conservation with a GDP/GTP exchange protein, Rab3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MADD
Applications Western Blot
Classification Recombinant Monoclonal
Clone JE63-62
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MADD
Gene Accession No. Q80U28, Q8WXG6
Gene Alias 9630059K23Rik; DENN; differentially expressed in normal and neoplastic cells; IG20; Insulinoma glucagonoma clone 20; insulinoma-glucagonoma protein 20; Kiaa0358; Madd; MAP kinase activating death domain; MAP kinase-activating death domain protein; MAP-kinase activating death domain; rab3 GDP/GTP exchange factor; Rab3 GDP/GTP exchange protein; Rab3 GEP; RAB3GEP
Gene Symbols MADD
Host Species Rabbit
Immunogen Synthetic peptide within human DENN aa 1,598-1,647/1,647.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228355, 8567
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.